Skip to Content
Merck
All Photos(1)

Key Documents

DWKBDA242P

Azlon Straight-sided Beakers

graduated, polypropylene, capacity 2000 mL

Synonym(s):

BDA242P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121803
NACRES:
NB.43

material

polypropylene

feature

graduated

packaging

pkg of 1 ea

manufacturer/tradename

Azlon

storage condition

room temperature

capacity

2000 mL

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Ideal for general laboratory use and management of liquid samples

Features and Benefits

Straight sided stackable

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service