CLS401392
Corning® Modified Impeller Assembly (for use with probes) for Magnetically-Driven Proculture® Spinner Flask
fits flask, 8 L
Synonym(s):
equipment replacement accessory
All Photos(1)
About This Item
UNSPSC Code:
41122104
NACRES:
NB.43
Recommended Products
material
stainless steel shaft
sterility
non-sterile
feature
fits flask 8 L
manufacturer/tradename
Corning 401392
Related Categories
General description
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, 8L Proculture Spinner Flasks. Narrower impeller paddle design allows use with probes to create a small bioreactor.
Legal Information
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)