BR782300
BRAND® strip plates
size 96 wells, 12xF8, MB, dividable: no
Synonym(s):
BRANDplates®, Microplates, Plates, Strip Plates
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
polystyrene
transparent wells
transparent wells
size:
96 wells
binding type:
medium binding surface
feature:
12xF8, MB
pureGrade
pureGrade
Recommended Products
material
polystyrene
transparent wells
feature
12xF8, MB
pureGrade
packaging
pack of 100 ea (4 bags of 25)
manufacturer/tradename
BRAND 782300
size
96 wells
binding type
medium binding surface
Related Categories
General description
BRANDplates® microtitration plates offer ideal visual properties and are produced according to ANSI/SLAS standards. They offer excellent flexibility. Only as many cavities as necessary are used. Two versions are available :Frame without grate:for processing 12 strips of 8 wells. Individual strips can be removed from the frame and re-inserted. Strips are not separable.Frame with grate :also 12 strips of 8 wells. Individual strips can be removed from and re-inserted into the plate. Strips can be divided in up to 8 individual tubes and inserted individually into the frame plate.They are available as 96-well standard plates as well as strip plates with pureGrade surfaces – for greater flexibility and optimal results.pureGrade medium binding capacity (medium-binding), somewhat more hydrophobic than immunoGradeCERTIFIED LIFE SCIENCE QUALITY: Virgin raw materials, Manufactured under controlled room conditions, Intensive in-process controls, Visual inspections, Batch management, Final product inspection, Proven functionality
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)