BR781390
BRAND® sealing film for microplates
self-adhesive, suitable for (PCR), polypropylene
Synonym(s):
Accessories, PCR, Sealing film
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
Recommended Products
material
polyester film
polypropylene
sterility
non-sterile
feature
piercable
self-adhesive
packaging
pack of 100 ea
manufacturer/tradename
BRAND 781390
technique(s)
PCR: suitable
suitability
suitable for (PCR)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
BRAND® offers film sheets, which can be applied on the plates and removed without the use of expensive equipment or can be applied with a sealing paddle to ensure secure and uniform attachment of the film to the plate. The source material is always valuable and the volume always low. This film is suitable for PCR, ELISA, EIA and other optical applications. It is transparent, enabling visual inspection of samples. Two tabs make it easy to handle, and the strong adhesive minimizes evaporation, while ensuring that the film remains securely in place on all plate types.
Features and Benefits
- Single film
- Temperature stability up to 120°C.
- Tight seal for minimizing evaporation.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service