BR136325
BRAND® stirring rod
high-density polyethylene (with steel core), L × D 300 mm × 6 mm
Synonym(s):
PTFE, Stirring rods
All Photos(3)
About This Item
UNSPSC Code:
41121802
NACRES:
NB.72
Recommended Products
material
high-density polyethylene (with steel core)
description
with steel core
packaging
pack of 1 ea
manufacturer/tradename
BRAND 136325
L × D
300 mm × 6 mm
Related Categories
General description
The BRAND® stirring rod is designed to be bendable, allowing for customization based on specific applications. The permanent bend ensures that the rod maintains its shape during use, providing flexibility for stirring in various vessel shapes and sizes. This innovative design enhances the efficiency of mixing and blending processes, making it suitable for a wide range of laboratory tasks.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)