Key Documents
BAF131600005
SP Bel-Art® disposal bag
width × H × thickness 12 in. (300 mm) × 24 in. (610 mm) × 0.04 mm (1.5 mil), translucent polypropylene, pack of 100 ea
Synonym(s):
biohaz waste bag, biohazard disposal bags, waste bag
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
material
translucent polypropylene
feature
label (Without Biohazard Warning Label or Sterilization Indicator Patch)
packaging
pack of 100 ea
manufacturer/tradename
Bel-Art F13160-0005
capacity
3.8-11.4 L (1-3 gal)
width × H × thickness
12 in. (300 mm) × 24 in. (610 mm) × 0.04 mm (1.5 mil)
General description
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.
Features and Benefits
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.
  • Transparent polypropylene; 0.03mm (1.5 mil) thick
  • Pass ASTM
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)