Skip to Content
Merck
All Photos(1)

Key Documents

ALD00532

Sigma-Aldrich

Serine Hydrolase Inhibitor-11

95%

Synonym(s):

Piperidin-1-yl(1H-pyrazol-1-yl)methanone, SHI-11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O
CAS Number:
Molecular Weight:
179.22
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

SMILES string

O=C(N1CCCCC1)N2N=CC=C2

InChI

1S/C9H13N3O/c13-9(12-8-4-5-10-12)11-6-2-1-3-7-11/h4-5,8H,1-3,6-7H2

InChI key

RVUWRYIKKXZWFA-UHFFFAOYSA-N

Application

Serine hydrolase inhibitors are small molecule fragments that react in vitro with the serine hydrolase class of proteins. They are amenable to further modification to create novel inhibitors of this class of protein.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katerina Otrubova et al.
Bioorganic & medicinal chemistry letters, 24(16), 3807-3813 (2014-07-20)
Two libraries of modestly reactive ureas containing either electron-deficient acyl anilines or acyl pyrazoles were prepared and are reported as screening libraries for candidate serine hydrolase inhibitors. Within each library is a small but powerful subset of compounds that serve

Related Content

Boger Lab utilizes nature's solutions in complex natural products, extending them through rational design for improved biological outcomes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service