Skip to Content
Merck
All Photos(3)

Key Documents

929662

Sigma-Aldrich

ElectroGreen®

greener alternative

NMP substitute for electronics – type 2, bio-sourced

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NA.05

grade

for electronic purposes

Quality Level

vapor pressure

0.05 mmHg ( 25 °C)

description

Relative Evaporation rate: <0.02

Hansen Solubility Parameters: SPd = 9.0; SPp = 6.8; SPh = 4.6.

Assay

≥99% (GC)

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

impurities

≤0.05% Acidity (as lactic acid)
≤0.2 wt. % Water (Karl Fischer)
≤1 ppm As, Cr, Cd, Cu, Hg, Mn, Ni, Pb, Zn, trace (each)

evapn. residue

≤0.05%

color

clear

viscosity

32.9  cP(20 °C)

bp

189 °C (initial)

density

0.996 at 25 °C (specific gravity)

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

This ElectroGreen® solvent is made of bio-based (corn/beets-derived) safer alternatives to replace synthetic NMP fossil-based solvent with specifications tailored for electronics and energy applications. All our ElectroGreen® solvents line are derived from a renewable resource (verified through C14 ASTMD6866-16 testing). We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Click here for more information.

This solvent blend consists of Dimethyl sulphoxide 50-70%; N,N-Dimethyloctanamide 20-30%; N,N-Dimethyldecan-1-amide 10-20%.

Application

N-Methyl Pyrrolidone (NMP)’s largest use in electronics is in cleaning, surface preparation applications and ink formulations. For coatings formulators looking to replace petroleum-based solvents with a bio-based, sustainable alternative, NMP substitute – type 2, bio-sourced is a solvent blend ideal replacement for NMP in a wide array of ink formulations and cleaning applications.

Storage and Stability

To ensure low water content, handle under inert conditions. Product should be refrigerated upon receipt.

Legal Information

ElectroGreen is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F

Flash Point(C)

85 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Carbon-based Sustainable Organic Electronics (SOE) limit the use of critical elements and biodegrade at their end-of-life. This review offers insight on how structural and energy disorder in these materials influence device performance and includes evaluations of various transport models and their limitations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service