914681
Aluminum oxide milled nanofibers alpha alumina
Synonym(s):
Al2O3, Nanofibrous inorganic powder
About This Item
Recommended Products
description
Thermal Conductivity 30 Wm-1K-1
Quality Level
form
nanofiber
powder (fluffy)
fiber diam. × L
300-900 nm ± 100 nm × 2-10 μm , Structure: Polycystalline nanofibers
surface area
30-50 m2/g
color
white
primary crystallite size
(alpha)
conductivity
<10^-16 S/cm (Electrical)
mp
2072 °C
SMILES string
[Al+3].[Al+3].[O-2].[O-2].[O-2]
InChI
1S/2Al.3O/q2*+3;3*-2
InChI key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
Application
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service