Skip to Content
Merck
All Photos(3)

Key Documents

903426

Sigma-Aldrich

Ru(p-CF3-bpy)3 (BF4)2

≥95%

Synonym(s):

Tris(2,2′-(pCF3)bipyridine)ruthenium(II) tetrafluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CLP 117,000

CLP 117,000


Estimated to ship onApril 11, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
CLP 117,000

About This Item

Empirical Formula (Hill Notation):
C36H18B2F26N6Ru
Molecular Weight:
1151.22
UNSPSC Code:
12352101

CLP 117,000


Estimated to ship onApril 11, 2025


Request a Bulk Order

Assay

≥95%

form

powder or crystals

reaction suitability

core: ruthenium
reagent type: catalyst
reaction type: Photocatalysis

mp

>300 °C

Application

Ru(p-CF3-bpy)3 (BF4)2 is a versatile photocatalyst that can be used for a variety of synthetic transformations.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Dear Sir can you clarify me that this chemical will be suitable to use fabricate OLED devices. please clarify.

    1 answer
    1. This product has not been validated for suitability for OLED fabrication. The end-user must determine the suitability. Similar Ruthenium bipyridine complex materials have been published for OLED fabrication.

      Please see the link below to review a publication that may be helpful:
      https://pubs.acs.org/doi/10.1021/ja000666t

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service