Skip to Content
Merck
All Photos(1)

Key Documents

901579

Sigma-Aldrich

t-Boc-N-amido-PEG1-CH2CO2H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO5
CAS Number:
Molecular Weight:
219.23
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: cross-linking reagent

functional group

Boc
amine
carboxylic acid

storage temp.

−20°C

SMILES string

O=C(O)COCCNC(OC(C)(C)C)=O

InChI

1S/C9H17NO5/c1-9(2,3)15-8(13)10-4-5-14-6-7(11)12/h4-6H2,1-3H3,(H,10,13)(H,11,12)

InChI key

UPBQMAHYLWJGDW-UHFFFAOYSA-N

Application

This is a crosslinker with a t-Boc protected amine on one end and a carboxyl group on the other end. The compound contains a single PEG unit to help improve solubility
t-Boc-N-amido-PEG1-CH2CO2H is a heterobifunctional, PEGylated crosslinker featuring a Boc-protected amine at one end and a carboxyl group at the other. The hydrophillic PEG linker facilitates solubility in biological applications. t-Boc-N-amido-PEG1-CH2CO2H can be used for bioconjugation or as a building block for synthesis of small molecules, conjugates of small molecules and/or biomolecules, or other tool compounds for chemical biology and medicinal chemistry that require ligation. Examples of applications include its synthetic incorporation into antibody-drug conjugates or proteolysis-targeting chimeras (PROTAC® molecules) for targeted protein degradation.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service