Skip to Content
Merck
All Photos(1)

Key Documents

809748

Sigma-Aldrich

(24R)-24,25-Dihydroxyvitamin D3 solution

100 μg/mL in ethanol, ≥97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O3
Molecular Weight:
416.64
EC Number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

Quality Level

Assay

≥95% (R Isomer)
≥97% (CP)

form

solution

concentration

100 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

storage temp.

−20°C

SMILES string

C=C1CC[C@H](O)C/C1=C/C=C2C(CC[C@@H]3[C@@H](CC[C@@H](O)C(O)(C)C)C)[C@]3(C)CCC/2

InChI

1S/C27H44O3/c1-18-8-12-22(28)17-21(18)11-10-20-7-6-16-27(5)23(13-14-24(20)27)19(2)9-15-25(29)26(3,4)30/h10-11,19,22-25,28-30H,1,6-9,12-17H2,2-5H3/b20-10+,21-11-/t19-,22+,23-,24?,25-,27-/m1/s1

InChI key

FCKJYANJHNLEEP-AOWQBJNISA-N

General description

(24R)-24,25-Dihydroxyvitamin D3 is a metabolite of vitamin D3. It is known to increase bone volume and mechanical strength.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

24R, 25-dihydroxyvitamin D3: an essential vitamin D3 metabolite for both normal bone integrity and healing of tibial fracture in chicks
Seo E, et al.
Endocrinology, 138(9), 3864-3872 (1997)
Analysis of C-3 epimerization in (24R)-24, 25-dihydroxyvitamin D3 catalyzed by hydroxysteroid dehydrogenase
Higashi T, et al.
Journal of Pharmaceutical and Biomedical Analysis, 36(3), 429-436 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service