Skip to Content
Merck
All Photos(1)

Key Documents

809063

Sigma-Aldrich

Sodium 1-Phenoxy-methanesulfinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NaO3S
Molecular Weight:
194.18
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

powder or crystals

Quality Level

reaction suitability

reaction type: C-C Bond Formation
reaction type: C-H Activation
reagent type: catalyst

functional group

phenoxy
sulfinic acid

SMILES string

O=S(O[Na])COC1=CC=CC=C1

InChI

1S/C7H8O3S.Na/c8-11(9)6-10-7-4-2-1-3-5-7;/h1-5H,6H2,(H,8,9);/q;+1/p-1

InChI key

KITYKPJDXMLWTO-UHFFFAOYSA-M

General description

Sodium 1-Phenoxy-methanesulfinate is a sulfinate reagent bearing a phenoxy moiety.

Application

The following Baran Sulfinate is a part of a toolbox of diversification reagents which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.

Legal Information

Diversinates is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service