Skip to Content
Merck
All Photos(2)

Key Documents

772194

Sigma-Aldrich

Phenyl α-D-thiomannopyranoside

97%

Synonym(s):

Phenyl 1-thio-α-D-mannopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16O5S
CAS Number:
Molecular Weight:
272.32
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

optical activity

[α]22/D +254°, c = 0.5% in methanol

mp

126-131 °C

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@H](O)[C@@H](SC2=CC=CC=C2)O[C@H](CO)[C@H]1O

InChI

1S/C12H16O5S/c13-6-8-9(14)10(15)11(16)12(17-8)18-7-4-2-1-3-5-7/h1-5,8-16H,6H2/t8-,9-,10+,11+,12-/m1/s1

InChI key

OVLYAISOYPJBLU-LDMBFOFVSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service