741531
3-Bromo-1,2-diamino-5-fluorobenzene
97%
Synonym(s):
3-Bromo-5-fluoro-1,2-phenylenediamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
form
solid
mp
70-75 °C
storage temp.
2-8°C
SMILES string
Nc1cc(F)cc(Br)c1N
InChI
1S/C6H6BrFN2/c7-4-1-3(8)2-5(9)6(4)10/h1-2H,9-10H2
InChI key
VCINLTLIJKUOQC-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service