736716
Potassium hexacyanoferrate(II)-13C6 trihydrate
99 atom % 13C, 98% (CP)
Synonym(s):
Potassium ferrocyanide-13C6
About This Item
Recommended Products
isotopic purity
99 atom % 13C
Assay
98% (CP)
form
solid
mass shift
M+6
SMILES string
O.O.O.[K+].[K+].[K+].[K+].N#[13C][Fe-4]([13C]#N)([13C]#N)([13C]#N)([13C]#N)[13C]#N
InChI
1S/6CN.Fe.4K.3H2O/c6*1-2;;;;;;;;/h;;;;;;;;;;;3*1H2/q;;;;;;-4;4*+1;;;/i6*1+1;;;;;;;;
InChI key
NRYOVBQTVIWDNU-NSNQEQAISA-N
Packaging
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service