Skip to Content
Merck
All Photos(1)

Key Documents

718351

Sigma-Aldrich

S,S′-[1,4-Phenylenebis(2,1-ethynediyl-4,1-phenylene)]bis(thioacetate)

97% (HPLC)

Synonym(s):

Thioacetic acid S-[4-[4-(4-acetylsulfanylphenylethynyl)-phenylethynyl]-phenyl] ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H18O2S2
CAS Number:
Molecular Weight:
426.55
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97% (HPLC)

form

powder

mp

179-189 °C

storage temp.

2-8°C

SMILES string

CC(=O)Sc1ccc(cc1)C#Cc2ccc(cc2)C#Cc3ccc(SC(C)=O)cc3

InChI

1S/C26H18O2S2/c1-19(27)29-25-15-11-23(12-16-25)9-7-21-3-5-22(6-4-21)8-10-24-13-17-26(18-14-24)30-20(2)28/h3-6,11-18H,1-2H3

InChI key

HTBKJDXAIPKGEY-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Hydrolyzing agents deprotect protected thiol derivatives, obtaining unstable free thiols during complex compound synthesis for self-assembled monolayers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service