Skip to Content
Merck
All Photos(1)

Documents

715492

Sigma-Aldrich

4-(Dimethylcarbamoyl)phenylboronic acid

≥94%

Synonym(s):

(4-(Dimethylaminocarbonyl)phenyl)boronic acid, 4-(N,N-Dimethylaminocarbonyl)phenylboronic acid, 4-(Dimethylcarbamoyl)benzeneboronic acid, 4-Dimethylcarbamoylphenylboronic acid, [4-(N,N-Dimethylaminocarbonyl)phenyl]boronic acid, [4-(N,N-Dimethylcarbamoyl)phenyl]boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12BNO3
CAS Number:
Molecular Weight:
193.01
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥94%

form

solid

mp

141-145 °C

storage temp.

2-8°C

SMILES string

CN(C)C(=O)c1ccc(cc1)B(O)O

InChI

1S/C9H12BNO3/c1-11(2)9(12)7-3-5-8(6-4-7)10(13)14/h3-6,13-14H,1-2H3

InChI key

QJYYVSIRDJVQJW-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride

Application

Reactant for preparation of:
  • Selective glucocorticoid receptor agonists
  • (thienopyridine)carboxamides as checkpoint 1 kinase (CHK1) inhibitors
  • [(pyrrolo[2,3-b]pyridinyl)methylene]hydroxybenzofuranones as ATP-competitive inhibitors of the mammalian target of rapamycin
  • Pyrrolopyridines as inhibitors of insulin-like growth factor-1 receptor tyrosine kinase

Reactant for:
  • Copper-mediated N-arylation of quinazolinediones

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service