684511
Methyl 16-bromohexadecanoate
97%
Synonym(s):
16-bromohexadecanoic acid, methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H33BrO2
CAS Number:
Molecular Weight:
349.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
mp
30-34 °C
functional group
bromo
ester
SMILES string
COC(=O)CCCCCCCCCCCCCCCBr
InChI
1S/C17H33BrO2/c1-20-17(19)15-13-11-9-7-5-3-2-4-6-8-10-12-14-16-18/h2-16H2,1H3
InChI key
ZYTQDEJMRYPSHE-UHFFFAOYSA-N
Application
Omega-functionalized long chain protected carboxylic acid. Utility: building block for self-assembled monolayer molecules.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mukesh K Pandey et al.
Nuclear medicine and biology, 80-81, 13-23 (2019-11-24)
The objectives of the present work were to optimize and validate the synthesis and stability of 14(R,S)-[18F]fluoro-6-thia-heptadecanoic acid ([18F]FTHA) and 16-[18F]fluoro-4-thia-palmitic acid ([18F]FTP) under cGMP conditions for clinical applications. Benzyl-14-(R,S)-tosyloxy-6-thiaheptadecanoate and methyl 16-bromo-4-thia-palmitate were used as precursors for the synthesis
Shane Pawsey et al.
Journal of the American Chemical Society, 125(14), 4174-4184 (2003-04-03)
The structures formed by the adsorption of carboxyalkylphosphonic acids on metal oxides were investigated by (1)H fast magic angle spinning (MAS), heteronuclear correlation (HETCOR), and (1)H double-quantum (DQ) MAS solid-state NMR experiments. The diacids HO(2)C(CH(2))(n)PO(3)H(2) (n = 2, 3, 11
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service