All Photos(1)
About This Item
Linear Formula:
IC6H4COCH3
CAS Number:
Molecular Weight:
246.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
solid
bp
107 °C/1 mmHg
density
1.767 g/mL at 25 °C
functional group
iodo
ketone
SMILES string
CC(=O)c1cccc(I)c1
InChI
1S/C8H7IO/c1-6(10)7-3-2-4-8(9)5-7/h2-5H,1H3
InChI key
IWLHOUBDKCKJJQ-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
>230.0 °F
Flash Point(C)
> 110 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service