Skip to Content
Merck
All Photos(1)

Key Documents

608092

Sigma-Aldrich

L-Isoleucine-13C6,15N

98 atom % 15N, 98 atom % 13C, 95% (CP)

Synonym(s):

SILAC Amino Acid, (2S,3S)-2-Amino-3-methylpentanoic acid-13C6,15N, 13C and 15N Labeled L-isoleucine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CLP 931,000
250 MG
CLP 1,390,000

CLP 931,000


Estimated to ship onMay 16, 2025


Request more information

Select a Size

Change View
100 MG
CLP 931,000
250 MG
CLP 1,390,000

About This Item

Linear Formula:
13C2H513CH(13CH3)13CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
138.12
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

CLP 931,000


Estimated to ship onMay 16, 2025


Request more information

isotopic purity

98 atom % 13C
98 atom % 15N

Quality Level

Assay

95% (CP)

form

solid

optical activity

[α]25/D +40.0°, c = 2 in 5 M HCl

technique(s)

bio NMR: suitable
protein expression: suitable

mp

168-170 °C (lit.)

mass shift

M+7

SMILES string

[13CH3][13CH]([13CH3])[13CH2][13C@H]([15NH2])[13C](O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1

InChI key

ROHFNLRQFUQHCH-HUEJSTCGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

L-isoleucine is an essential amino acid that is synthesized from threonine. It is a branched-chain hydrophobic amino acid. L-isoleucine is an isomer of L-leucine.

Application

L-Isoleucine-13C6,15N is used in the immunocapture isotope dilution mass spectrometry (IC-IDMS) to evaluate the suitability of the underlying monoclonal andpolyclonal antibodies (mAbs and pAbs) for new vaccine quantification.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carrie L Pierce et al.
Vaccine, 35(37), 5011-5018 (2017-08-05)
As a result of recent advances in mass spectrometry-based protein quantitation methods, these techniques are now poised to play a critical role in rapid formulation of pandemic influenza vaccines. Analytical techniques that have been developed and validated on seasonal influenza

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service