Skip to Content
Merck
All Photos(1)

Key Documents

604801

Sigma-Aldrich

L-Proline-13C5

99 atom % 13C, 99% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CLP 1,760,000

CLP 1,760,000


Estimated to ship onApril 19, 2025


Request more information

Select a Size

Change View
100 MG
CLP 1,760,000

About This Item

Empirical Formula (Hill Notation):
13C5H9NO2
CAS Number:
Molecular Weight:
120.09
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

CLP 1,760,000


Estimated to ship onApril 19, 2025


Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

form

solid

optical activity

[α]25/D -86.0°, c = 2 in H2O

mp

228 °C (dec.) (lit.)

mass shift

M+5

storage temp.

room temp

SMILES string

O[13C](=O)[13C@@H]1[13CH2][13CH2][13CH2]N1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1/i1+1,2+1,3+1,4+1,5+1

InChI key

ONIBWKKTOPOVIA-JRGPAWSWSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service