Skip to Content
Merck
All Photos(2)

Key Documents

570400

Sigma-Aldrich

Disperse Red 1 acrylate

95%

Synonym(s):

2-[N-ethyl-4-[(4-nitrophenyl)diazenyl]anilino]ethyl prop-2-enoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4N=NC6H4N(C2H5)CH2CH2OCOCCH=CH2
CAS Number:
Molecular Weight:
368.39
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

mp

85-104 °C (dec.) (lit.)

λmax

492 nm

storage temp.

2-8°C

SMILES string

CCN(CCOC(=O)C=C)c1ccc(cc1)\N=N\c2ccc(cc2)[N+]([O-])=O

InChI

1S/C19H20N4O4/c1-3-19(24)27-14-13-22(4-2)17-9-5-15(6-10-17)20-21-16-7-11-18(12-8-16)23(25)26/h3,5-12H,1,4,13-14H2,2H3/b21-20+

InChI key

MOUZYIAKMYUILJ-QZQOTICOSA-N

General description

NLO material

Application

  • This research uses Disperse Red 1 acrylate in liquid crystal monomers to achieve specific voxel orientations, impacting materials science in robotics and sensors (Guo et al., 2021).
  • The study investigates ink formulations that include Disperse Red 1 acrylate for textile printing applications, demonstrating advances in fabric processing (Song et al., 2018).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Li, Najun;
Polymer, 50, 428-433 (2009)
Miki Ishifuji et al.
Journal of the American Chemical Society, 131(12), 4418-4424 (2009-03-12)
Flexible design of hybrid polymer nanoassemblies consisting of nonlinear optical (NLO) polymer nanosheets and gold nanoparticle alignment was done to elucidate near-field effects of localized surface plasmon (LSP) coupling, which was generated from coupled gold nanoparticles, on enhanced second harmonic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service