Skip to Content
Merck
All Photos(2)

Key Documents

544914

Sigma-Aldrich

IR-775 chloride

Dye content ~90 %

Synonym(s):

2-[2-[2-Chloro-3-[2-(1,3-dihydro-1,3,3-trimethyl-2H-indol-2-ylidene)-ethylidene]-1-cyclohexen-1-yl]-ethenyl]-1,3,3-trimethyl-3H-indolium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H36Cl2N2
CAS Number:
Molecular Weight:
519.55
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

composition

Dye content, ~90%

mp

208-215 °C (lit.)

λmax

775 nm

SMILES string

[Cl-].CN1c2ccccc2C(C)(C)\C1=C\C=C3/CCCC(\C=C\C4=[N+](C)c5ccccc5C4(C)C)=C3Cl

InChI

1S/C32H36ClN2.ClH/c1-31(2)24-14-7-9-16-26(24)34(5)28(31)20-18-22-12-11-13-23(30(22)33)19-21-29-32(3,4)25-15-8-10-17-27(25)35(29)6;/h7-10,14-21H,11-13H2,1-6H3;1H/q+1;/p-1

InChI key

BPSIJFMUSNMMAL-UHFFFAOYSA-M

Features and Benefits

Solvent soluble, laser and near-IR dye

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service