470317
2,5-Thiophenediylbisboronic acid
≥95.0%
Synonym(s):
2,5-Thiophenediboronic acid
About This Item
Recommended Products
Quality Level
Assay
≥95.0%
mp
250 °C (dec.) (lit.)
SMILES string
OB(O)c1ccc(s1)B(O)O
InChI
1S/C4H6B2O4S/c7-5(8)3-1-2-4(11-3)6(9)10/h1-2,7-10H
InChI key
SKFDVFMUXZWWOW-UHFFFAOYSA-N
Application
- As a reactant in the metal catalyzed Suzuki cross coupling reactions.
- To prepare 2,5-bis(2-pyridyl)thiophene, a thiophene based ligand which is used in the design of fluorescent mercury-sensor.
- To fabricate graphene-based biosensors for the detection of glucose and Escherichia coli in a complex medium.
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service