Skip to Content
Merck
All Photos(1)

Key Documents

434124

Sigma-Aldrich

Poly(4-chlorostyrene)

average Mw ~75,000 by GPC, powder

Synonym(s):

p -Chlorostyrene polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH(C6H4Cl)-]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

mol wt

average Mw ~75,000 by GPC

transition temp

Tg 106 °C

SMILES string

Clc1ccc(C=C)cc1

InChI key

KTZVZZJJVJQZHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service