Skip to Content
Merck
All Photos(2)

Key Documents

376264

Sigma-Aldrich

Trimethyl(propargyl)silane

contains 500 ppm BHT as stabilizer

Synonym(s):

2-Propynyl-trimethylsilane, 3-(Trimethylsilyl)propyne

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCH2Si(CH3)3
CAS Number:
Molecular Weight:
112.24
Beilstein:
2036316
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

contains

500 ppm BHT as stabilizer
≤20% trimethylallenylsilane (in equilibrium)

refractive index

n20/D 1.413 (lit.)

bp

91-93 °C (lit.)

density

0.753 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

C[Si](C)(C)CC#C

InChI

1S/C6H12Si/c1-5-6-7(2,3)4/h1H,6H2,2-4H3

InChI key

ULYLMHUHFUQKOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethyl(propargyl)silane (2-Propynyl-trimethylsilane) is a clear colorless liquid and contains 500ppm of butylated hydroxytoluene (BHT) as stabilizer. It undergoes GaCl3-catalyzed dehydrogenation reaction with aldimines to afford 4-methylquinoline.

Application

Trimethyl(propargyl)silane is suitable for use in the synthesis of diverse end-functionalized (ω-carboxyl, ω-hydroxy, ω-methyl-vinyl, ω-trimethylsilane, and ω-glycidyl-ether) via ′′click′′ reaction. It may be used in the synthesis of γ-allenyl-GABA (γ-aminobutyric acid), via Lewis acid mediated reaction with ω-ethoxy lactams.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

23.0 °F - closed cup

Flash Point(C)

-5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GaCl3-catalyzed [4+2] annulations of allyltrimethylsilane and trimethyl (propargyl) silane with aldimines.
Hirashita T, et al.
Tetrahedron Letters, 48(31), 5421-5424 (2007)
Lewis acid induced reactions of propargyl trimethyl silane with ?-ethoxy lactams synthesis of ?-allenyl-GABA.
Hiemstra H, et al.
Tetrahedron Letters, 25(29), 3115-3118 (1984)
High yield synthesis of diverse well-defined end-functionalized polymers by combination of anionic polymerization and ?click? chemistry.
Ho C-C, et al.
Journal of Applied Polymer Science, 111(3), 1571-1580 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service