367656
1,1,1,3,3,3-Hexafluoroisopropyl acrylate
99%
Synonym(s):
HFiPA
About This Item
Recommended Products
vapor pressure
1.22 psi ( 20 °C)
Assay
99%
form
liquid
refractive index
n20/D 1.319 (lit.)
density
1.33 g/mL at 25 °C (lit.)
SMILES string
FC(F)(F)C(OC(=O)C=C)C(F)(F)F
InChI
1S/C6H4F6O2/c1-2-3(13)14-4(5(7,8)9)6(10,11)12/h2,4H,1H2
InChI key
MNSWITGNWZSAMC-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
50.0 °F - closed cup
Flash Point(C)
10 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service