Recommended Products
Assay
98%
form
liquid crystal
SMILES string
CCCCCCCc1ccc(cc1)-c2ccc(cc2)C#N
InChI
1S/C20H23N/c1-2-3-4-5-6-7-17-8-12-19(13-9-17)20-14-10-18(16-21)11-15-20/h8-15H,2-7H2,1H3
InChI key
ZGOWXOZNUNZPAV-UHFFFAOYSA-N
Related Categories
General description
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 4
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service