Skip to Content
Merck
All Photos(1)

Key Documents

329339

Sigma-Aldrich

1,4-Dichlorobenzene-d4

98 atom % D, 98% (CP)

Synonym(s):

Tetradeutero-1,4-dichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D4Cl2
CAS Number:
Molecular Weight:
151.03
Beilstein:
2047072
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

bp

173 °C (lit.)

mp

52-54 °C (lit.)

density

1.274 g/mL at 25 °C

application(s)

environmental

mass shift

M+4

storage temp.

2-8°C

SMILES string

[2H]c1c([2H])c(Cl)c([2H])c([2H])c1Cl

InChI

1S/C6H4Cl2/c7-5-1-2-6(8)4-3-5/h1-4H/i1D,2D,3D,4D

InChI key

OCJBOOLMMGQPQU-RHQRLBAQSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Célia A Alves et al.
Environmental science and pollution research international, 22(15), 11526-11542 (2015-04-02)
The main purpose of this work was to evaluate the chemical composition of particulate matter (PM) emitted by eight different light-duty vehicles. Exhaust samples from petrol and diesel cars (Euro 3 to Euro 5) were collected in a chassis dynamometer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service