All Photos(1)
About This Item
Linear Formula:
CH3S(CH2)4OH
CAS Number:
Molecular Weight:
120.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.487 (lit.)
bp
95-98 °C/20 mmHg (lit.)
density
0.993 g/mL at 25 °C (lit.)
functional group
hydroxyl
thioether
SMILES string
CSCCCCO
InChI
1S/C5H12OS/c1-7-5-3-2-4-6/h6H,2-5H2,1H3
InChI key
JNTVUHZXIJFHAU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
215.6 °F - closed cup
Flash Point(C)
102 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service