317810
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate
99%
Synonym(s):
Ethylenediaminetetraacetic acid disodium magnesium salt, Magnesium disodium ethylenediaminetetraacetate hydrate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
(NaOOCCH2)2NCH2CH2N(CH2COO)2Mg · xH2O
Molecular Weight:
358.50 (anhydrous basis)
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
functional group
amine
SMILES string
[Na+].[Na+].[H]O[H].[O-]C(=O)CN(CCN1CC(=O)O[Mg]OC(=O)C1)CC([O-])=O
InChI
1S/C10H16N2O8.Mg.2Na.H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;1H2/q;+2;2*+1;/p-4
InChI key
IFRDBGYJSLDMHQ-UHFFFAOYSA-J
Application
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate (disodium magnesium EDTA) can be used as:
- A chelating agent in various fields that binds a wide range of polyvalent cations, including calcium.
- A nitrogen precursor salt to synthesize nitrogen-doped porous carbon material via direct pyrolysis for supercapacitor applications.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service