Skip to Content
Merck
All Photos(1)

Documents

299243

Sigma-Aldrich

Phthalimide-15N potassium salt

98 atom % 15N

Synonym(s):

1,3-Dihydro-1,3-dioxoisoindole-15N, Potassium phthalimide-15N

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4K15NO2
CAS Number:
Molecular Weight:
186.21
Beilstein:
3639404
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

Quality Level

form

solid

mp

>300 °C (lit.)

mass shift

M+1

SMILES string

[K][15N]1C(=O)c2ccccc2C1=O

InChI

1S/C8H5NO2.K/c10-7-5-3-1-2-4-6(5)8(11)9-7;/h1-4H,(H,9,10,11);/q;+1/p-1/i9+1;

InChI key

FYRHIOVKTDQVFC-DLBIPZKSSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Labelled Compounds & Radiopharmaceuticals, 34, 663-663 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service