Skip to Content
Merck
All Photos(1)

Documents

290408

Sigma-Aldrich

3,4-Difluorophenol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3OH
CAS Number:
Molecular Weight:
130.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

85 °C/20 mmHg (lit.)

mp

34-38 °C (lit.)

SMILES string

Oc1ccc(F)c(F)c1

InChI

1S/C6H4F2O/c7-5-2-1-4(9)3-6(5)8/h1-3,9H

InChI key

BNPWVUJOPCGHIK-UHFFFAOYSA-N

General description

Transformation of 3,4-difluorophenol by Penicillium frequentans strain Bi 7/2, using phenol as a sole source of carbon and energy, has been reported.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Wunderwald et al.
Biodegradation, 8(6), 379-385 (1997-01-01)
The Penicillium frequentans strain Bi 7/2, using phenol as a sole source of carbon and energy, transformed the fluorinated phenols 2,3-, 2,4-, 2,5- and 3,4-difluorophenol rapidly. After growth on phenol, resting mycelia of the fungus converted the difluorophenols completely at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service