Skip to Content
Merck
All Photos(1)

Key Documents

279420

Sigma-Aldrich

Glycine-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
CLP 410,000

CLP 410,000


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
CLP 410,000

About This Item

Linear Formula:
H2NCH213CO2H
CAS Number:
Molecular Weight:
76.06
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.12

CLP 410,000


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

solid

mp

240 °C (dec.) (lit.)

mass shift

M+1

SMILES string

NC[13C](O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)/i2+1

InChI key

DHMQDGOQFOQNFH-VQEHIDDOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Krog Ingerslev et al.
Journal of proteome research, 14(8), 3095-3110 (2015-07-03)
The effects of increased intake of dietary fiber as either whole grain rye or as resistant starch compared to a typical low dietary fiber Western-style diet on the metabolomics responses were studied in gastrointestinal content and tissue, peripheral plasma, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service