Skip to Content
Merck
All Photos(1)

Key Documents

27826

Sigma-Aldrich

Poly[(R)-3-hydroxybutyric acid-co-(R)-3-hydroxyvaleric acid]

~9:1

Synonym(s):

Biopol, Copolymer of (R)-3-hydroxybutyric acid and (R)-3-hydroxyvaleric acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

SMILES string

O([C@H](CC)CC(=O)O[C@@H](CC(=O)O[C@H](CC)CC=O)C)C(=O)C[C@H](OC(=O)C[C@H](OC(=O)C[C@H](O)C)CC)C

InChI

1S/C27H44O12/c1-7-20(10-11-28)37-24(31)13-18(5)35-27(34)16-22(9-3)39-25(32)14-19(6)36-26(33)15-21(8-2)38-23(30)12-17(4)29/h11,17-22,29H,7-10,12-16H2,1-6H3/t17-,18-,19-,20-,21-,22-/m1/s1

InChI key

HFQOLOYVZLYECE-CGXUPHRESA-N

Looking for similar products? Visit Product Comparison Guide

General description

ratio (R)-3-hydroxybutyric acid : (R)-3-hydroxyvaleric acid ≥9:1 (NMR)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service