Skip to Content
Merck
All Photos(1)

Key Documents

274208

Sigma-Aldrich

Dichlorodiethylsilane

97%

Synonym(s):

Diethyldichlorosilane, Diethyldichlorosilicon

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2SiCl2
CAS Number:
Molecular Weight:
157.11
Beilstein:
605313
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.43 (lit.)

bp

125-131 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC[Si](Cl)(Cl)CC

InChI

1S/C4H10Cl2Si/c1-3-7(5,6)4-2/h3-4H2,1-2H3

InChI key

BYLOHCRAPOSXLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

79.0 °F

Flash Point(C)

26.1 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Peter Bieling et al.
The EMBO journal, 37(1), 102-121 (2017-11-17)
WASP-family proteins are known to promote assembly of branched actin networks by stimulating the filament-nucleating activity of the Arp2/3 complex. Here, we show that WASP-family proteins also function as polymerases that accelerate elongation of uncapped actin filaments. When clustered on
David Gräfe et al.
Nature communications, 9(1), 2788-2788 (2018-07-19)
Existing photoresists for 3D laser lithography that can be removed after development in a subtractive manner typically suffer from harsh cleavage conditions. Here, we report chemoselectively cleavable photoresists for 3D laser lithography based on silane crosslinkers, allowing the targeted degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service