Skip to Content
Merck
All Photos(1)

Key Documents

224359

Supelco

Amberlite IR120 Ion Exchange Resin

sodium form, 15-50 mesh

Synonym(s):

Amberlite IR120 Na+ form

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
CLP 45,300
1 KG
CLP 124,000

CLP 45,300


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
250 G
CLP 45,300
1 KG
CLP 124,000

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

CLP 45,300


Estimated to ship onMay 26, 2025


Request a Bulk Order

Product Name

Amberlite IR120 Na+ form, Na+-form

vapor density

>1 (vs air)

vapor pressure

17 mmHg ( 20 °C)

description

cross-linkage 8%

form

beads

crosslinking

8 % cross-linked

autoignition temp.

800 °F

moisture

45%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

quaternary ammonium

particle size

16-50 mesh

capacity

2.0 meq/mL by wetted bed volume

separation technique

anion exchange

SMILES string

[Na+].c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H10.C8H8.Na/c1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;/h3-8H,1-2H2;2-7H,1H2;/q;;+1

InChI key

ZWZSXPSZPITJQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amberlite IR120 is a strongly acidic cation exchange resin which may be used for the recovery of toxic metals.[1] It may be used in column chromatography. It is required for a ion-exchange resin to swell in the solvent for a higher rate of sorption and to exchange ions.[2]

Application

Amberlite IR120 Na+ form may be used in separation medium for TLC, paper chromatography and LPLC.
Strongly acidic cation exchange resin for decalicification.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion-exchange equilibria of Cu2+, Cd2+, Zn2+, and Na+ ions on the cationic exchanger Amberlite IR-120.
Valverde, Jose L., et al.
Journal of Chemical and Engineering Data, 46, 1404-1409 (2001)
Determination of intraparticle diffusivities of Na+/K+ in water and water/alcohol mixed solvents on a strong acid cation exchanger.
Rodriguez, Juan F., et al.
Industrial & Engineering Chemistry Research, 41, 3019-3027 (2002)

Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What is the particle size of the Amberlite® IR120 sodium form , Product 224359?

    1 answer
    1. The Amberlite IR120 Na screens at 16-50 mesh (nominal particle size of 300-1200 microns in diameter). The harmonic mean is 600-800 microns, with a uniformity coefficient maximum of 1.9.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  5. Can I get the Amberlite® IR120 sodium form, Product 224359, in the hydrogen form?

    1 answer
    1. Amberlite IR120 H is available from Sigma-Aldrich as Product Nos. 06428, 10322 and 216534; pricing and package size varies by country.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service