Skip to Content
Merck
All Photos(1)

Key Documents

221600

Sigma-Aldrich

4-Phenylsemicarbazide hydrochloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCONHNH2·HCl
CAS Number:
Molecular Weight:
187.63
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

215-217 °C (lit.)

solubility

alcohol: freely soluble
water: freely soluble

functional group

amine

SMILES string

Cl.NNC(=O)Nc1ccccc1

InChI

1S/C7H9N3O.ClH/c8-10-7(11)9-6-4-2-1-3-5-6;/h1-5H,8H2,(H2,9,10,11);1H

InChI key

BNXHWDWODIHJEH-UHFFFAOYSA-N

Application

4-Phenylsemicarbazide hydrochloride was used in the experiment to study whether i) [U-14C]palmitate and[2-14C]palmitate are metabolized to ketone bodies and ii) free glycerol was used as an energy source by the tissue from ketotic and normal sheep.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Taylor et al.
The Biochemical journal, 106(1), 289-292 (1968-01-01)
Labelled ketone bodies were produced readily from [U-(14)C]palmitate, [2-(14)C]palmitate and [1-(14)C]glycerol by sheep rumen-epithelial and liver tissues in vitro. On a tissue-nitrogen basis, both tissues had similar capacities for ketogenesis. Palmitate was a ketogenic substrate in both rumen-epithelial tissue and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service