Skip to Content
Merck
All Photos(1)

Documents

179272

Sigma-Aldrich

Diethyl ether

≥97.5%, contains BHT as stabilizer

Synonym(s):

Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
EC Number:
MDL number:
UNSPSC Code:
12352112
eCl@ss:
39021002
PubChem Substance ID:
NACRES:
NA.21

product name

Diethyl ether,

vapor density

2.6 (vs air)

Quality Level

vapor pressure

28.5 psi

Assay

≥97.5%

form

liquid

autoignition temp.

320 °F

contains

BHT as stabilizer

expl. lim.

36.5 %

impurities

2% ethyl alcohol

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl ether (DEE) is widely employed as solvent for Grignard synthesis. It has been reported as a hydroboration solvent. DEE is a promising compression-ignition fuel and its energy density is much higher than ethanol. It is also one of the components in a blend used in diesel fuel. It can also be used as a alternative of diesel fuel.

Application

Diethyl ether may be employed as a solvent in the following studies:
  • Oxidation of secondary alcohols into ketones.
  • Preparation of pentablock copolymer PAE-PCL-PEG-PCL-PAE [PAE = poly(β-amino ester), PEG = poly(ethylene glycol), PCL = poly(ε-caprolactone)].
  • Preparation of triblock copolymer of PCL-PEG-PCL.
  • Preparation of acrylated PCL-PEG-PCL.

Packaging

Packaged in seal-top can

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diethyl ether (DEE) as a renewable diesel fuel.
No. 972978. SAE technical paper (1997)
Dai P Huynh et al.
Biomaterials, 29(16), 2527-2534 (2008-03-11)
The concept of this research is using poly(beta-amino ester) (PAE) as a duo-functional group for synthesis of the novel sensitive injectable hydrogel for controlled drug/protein delivery. Firstly, PAE made of 1,4-butanediol diacrylate and 4,4'-trimethylene dipiperidine is used as a pH-sensitive
Oxidation of secondary alcohols in diethyl ether with aqueous chromic acid. Convenient procedure for the preparation of ketones in high epimeric purity.
Brown HC, et al.
The Journal of Organic Chemistry, 36(3), 387-390 (1971)
Eagleson M.
Concise Encyclopedia Chemistry, 471-471 (1994)
Juan E Camacho Londoño et al.
European heart journal, 36(33), 2257-2266 (2015-06-13)
Pathological cardiac hypertrophy is a major predictor for the development of cardiac diseases. It is associated with chronic neurohumoral stimulation and with altered cardiac Ca(2+) signalling in cardiomyocytes. TRPC proteins form agonist-induced cation channels, but their functional role for Ca(2+)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service