Skip to Content
Merck
All Photos(2)

Documents

127078

Sigma-Aldrich

N,N,N′,N′-Tetraethylethylenediamine

98%

Synonym(s):

TEEDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCH2CH2N(C2H5)2
CAS Number:
Molecular Weight:
172.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.4343 (lit.)

bp

189-192 °C (lit.)

density

0.808 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CCN(CC)CC

InChI

1S/C10H24N2/c1-5-11(6-2)9-10-12(7-3)8-4/h5-10H2,1-4H3

InChI key

DIHKMUNUGQVFES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N, N′,N′-Tetraethylethylenediamine (TEEDA) has been used in the synthesis of amine ligands using simple or double intramolecular dealkylation reaction. TEEDA exhibits selective β-lithiation during deprotonation with lithiumalkyls.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viktoria H Gessner et al.
Journal of the American Chemical Society, 130(44), 14412-14413 (2008-10-10)
TMEDA and its ethyl-substituted analogue, TEEDA, show a different behavior toward the deprotonation with lithiumalkyls. While TMEDA mainly undergoes alpha-lithiation of its methyl group, TEEDA shows selective beta-lithiation with following elimination of ethene. The crystal structure of the monomeric intermediate
M Esther Morilla et al.
Chemical communications (Cambridge, England), (17)(17), 1848-1849 (2002-09-26)
N,N, N',N'-Tetraethylethylenediamine undergoes simple or double intramolecular dealkylation reactions in the presence of the complexes [RuCl2(diene)]n (diene = cod, nbd) or trans-[RuCl2(diene)(morfoline)2 at 80 degrees C to afford chelating amine ligands which contains one or two N-H functionalities.
Lan-Fang Hu et al.
Molecules (Basel, Switzerland), 25(2) (2020-01-16)
The copolymerization of biorenewable succinic anhydride (SA) with propylene oxide (PO) is a promising way to synthesize biodegradable aliphatic polyesters. However, the catalytic systems for this reaction still deserve to be explored because the catalytic activity of the reported catalysts
Wei Xiao et al.
Biochimica et biophysica acta. General subjects, 1862(8), 1760-1769 (2018-05-12)
The inorganic core of the iron storage protein, ferritin, is recognized as being analogous to the poorly crystalline iron mineral, ferrihydrite (Fh). Fh is also abundant in soils where it is central to the redox cycling of particular soil contaminants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service