Principaux documents
CLS352350
Corning® Falcon® Cell Strainer For Use With 50ml Conical Tubes
pore size 70 μm, white Nylon, sterile
Synonyme(s) :
culture tube
About This Item
Code UNSPSC :
41104932
Nomenclature NACRES :
NB.13
Matériaux
white Nylon
Stérilité
sterile
Fabricant/nom de marque
Corning 352350
Dimension de pores
70 μm pore size
Application(s)
filtration
Informations légales
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.