Principaux documents
Z126055
Ace McLeod vacuum gauge
pressure 0-10 mmHg
About This Item
Code UNSPSC :
41112406
Nomenclature NACRES :
NB.79
Fabricant/nom de marque
Ace Glass 872807
Paramètres
0-10 mmHg pressure
Description générale
With interchangeable measuring tube. Mercury not included.