Principaux documents
Z118885
Spinbar® magnetic stirring flea assortment
pack of 12 ea
About This Item
Code UNSPSC :
41103806
Nomenclature NACRES :
NB.72
Matériaux
PTFE-coated
cylindrical PTFE-coated bar
Conditionnement
pack of 12 ea
Fabricant/nom de marque
Bel-Art F37186-0000
Description générale
1 of each of blue, red and yellow: 8 × 1.5 mm, 5 × 2 mm, 7 × 2 mm, 10 × 3 mm.
Informations légales
Spinbar is a registered trademark of SP Scienceware, a division of SP