Principaux documents
Z101133
Stainless steel 304 syringe needle
gauge 20, L 12 in.
Synonyme(s) :
Stainless Needle, Steel Syringe
About This Item
Code UNSPSC :
42142600
eCl@ss :
32070500
Nomenclature NACRES :
NB.03
Description
Standard Hub with 304 Stainless Steel Tubing
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Cadence Science 4188
Longueur
12 in.
Calibre
20
Description générale
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing