Principaux documents
SLW1516/10RED
Pyrex® Media-Lab Bottles, with high temperature cap and pouring ring, with printed trace code
capacity 1000 mL
Synonyme(s) :
Pyrex, SciLabware 1516/10RED
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Capacité:
1000 mL
Diam. × H:
101 mm × 225 mm
Matériaux:
PTFE seal
borosilicate glass bottle
red polyester cap (high temperature thermoplastic)
round bottle
white graduation
Caractéristiques:
graduations
with cap
with pouring ring
with printed trace code
Conditionnement:
pack of 10 ea
Matériaux
PTFE seal
borosilicate glass bottle
red polyester cap (high temperature thermoplastic)
round bottle
white graduation
Joint
Joints threaded neck (GL45)
Caractéristiques
graduations
with cap
with pouring ring
with printed trace code
Conditionnement
pack of 10 ea
Fabricant/nom de marque
SciLabware 1516/10RED
Capacité
1000 mL
Diam. × H
101 mm × 225 mm
Description générale
  • Bottles complete with thermoplastic polyester cap, PTFE sealing disc and pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Withstands hot air sterilisation up to 180 deg. C
  • PTFE sealing disc resists chemical attack
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
Informations légales
PYREX is a registered trademark of Corning, Inc.