Principaux documents
BR122528
BRAND® laboratory bottle, graduated with screw cap
capacity 50 mL
Synonyme(s) :
Bottles, Glass, Laboratory bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Capacité:
50 mL
Diam. × H:
46 mm × 91 mm
Matériaux:
borosilicate glass bottle
clear bottle
narrow-mouth bottle
polypropylene cap
round bottle
Stérilité:
non-sterile
Caractéristiques:
autoclavable
graduations
with cap
Conditionnement:
pack of 10 ea
Matériaux
borosilicate glass bottle
clear bottle
narrow-mouth bottle
polypropylene cap
round bottle
Joint
Joints threaded neck (GL32)
Stérilité
non-sterile
Caractéristiques
autoclavable
graduations
with cap
Conditionnement
pack of 10 ea
Fabricant/nom de marque
BRAND 122528
Capacité
50 mL
Diam. × H
46 mm × 91 mm
Description générale
Threaded to DIN ISO 4796. Studded base for stability and abrasion resistance. Graduated, with marking area. Screw cap and pouring ring PP. Autoclavable (121 °C).
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG