Merck
  • CDK-dependent phosphorylation of PHD1 on serine 130 alters its substrate preference in cells.
CDK-dependent phosphorylation of PHD1 on serine 130 alters its substrate preference in cells.
Journal of cell science (2015-12-09)
Brian Ortmann, Dalila Bensaddek, Sara Carvalhal, Sandra C Moser, Sharon Mudie, Eric R Griffis, Jason R Swedlow, Angus I Lamond, Sonia Rocha
ABSTRACT
PHD1 (also known as EGLN2) belongs to a family of prolyl hydroxylases (PHDs) that are involved in the control of the cellular response to hypoxia. PHD1 is also able to regulate mitotic progression through the regulation of the crucial centrosomal protein Cep192, establishing a link between the oxygen-sensing and the cell cycle machinery. Here, we demonstrate that PHD1 is phosphorylated by CDK2, CDK4 and CDK6 at S130. This phosphorylation fluctuates with the cell cycle and can be induced through oncogenic activation. Functionally, PHD1 phosphorylation leads to increased induction of hypoxia-inducible factor (HIF) protein levels and activity during hypoxia. PHD1 phosphorylation does not alter its intrinsic enzymatic activity, but instead decreases the interaction between PHD1 and HIF1α. Interestingly, although phosphorylation of PHD1 at S130 lowers its activity towards HIF1α, this modification increases the activity of PHD1 towards Cep192. These results establish a mechanism by which cell cycle mediators, such as CDKs, temporally control the activity of PHD1, directly altering the regulation of HIF1α and Cep192.
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
ChIPAb+ HDAC1 Antibody, rabbit polyclonal, from rabbit
Sigma-Aldrich
Anti-Phosphoserine Antibody, clone 4A4 (mouse IgG1), clone 4A4, Upstate®, from mouse