Merck
  • Clinical utility of acarbose, an alpha-glucosidase inhibitor in cardiometabolic disorders.
Clinical utility of acarbose, an alpha-glucosidase inhibitor in cardiometabolic disorders.
Current drug metabolism (2009-03-12)
Sho-ichi Yamagishi, Takanori Matsui, Seiji Ueda, Kei Fukami, Seiya Okuda
ABSTRACT
Diabetes is associated with an increase risk for cardiovascular disease (CVD). Recently, macrovascular complications of diabetes have been shown to start before the development of diabetes. Indeed, several clinical studies have confirmed the increased risk of CVD in patients with impaired glucose tolerance (IGT). Since postprandial hyperglycemia and insulin resistance are thought to play a central role in the development and progression of CVD in patients with IGT, amelioration of postprandial hyperglycemia as well as insulin resistance is a therapeutic target for the prevention of CVD in these high-risk patients. Acarbose, an alpha-glucosidase inhibitor, delays the absorption of carbohydrate from the small intestine, thereby reducing postprandial hyperglycemia. Further, recently, acarbose has been shown to improve insulin resistance in vivo. These findings suggest that acarbose is a promising metabolic modifier that could reduce the risk of CVD in patients with the metabolic syndrome. In this paper, we review the clinical utility of acarbose in various cardiometabolic disorders.
Product Description
Acarbose for peak identification, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Acarbose for identification, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acarbose, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acarbose, ≥95% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Acarbose, European Pharmacopoeia (EP) Reference Standard