Skip to Content
Merck
All Photos(1)

Documents

1700013

USP

Trypsin Recombinant Porcine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Trypsin porcine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

General description

Recombinant trypsin, a key raw material in biopharmaceutical manufacturing, is a serine protease that cleaves peptide chains mainly at the carboxyl end of the amino acids arginine and lysine. The amino acid sequence of recombinant trypsin is identical to that of trypsin from porcine pancreas, and the recombinant trypsin is produced by methods based on recombinant DNA technology in the yeast Pichia pastoris. Because of the recombinant production process, recombinant trypsin is free of chymotrypsin.

Trypsin Recombinant Porcine USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Trypsin Recombinant Porcine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Trypsin

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?89? ENZYMES USED AS ANCILLARY MATERIALS IN PHARMACEUTICAL MANUFACTURING
USPNF 2021 ISSUE 3: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(3) (2017)
ENZYMES USED AS ANCILLARY MATERIALS IN PHARMACEUTICAL MANUFACTURING
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(3), 6521-6521 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service